missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other SPA-Cys Long Protein

A cDNA sequence encoding the SPA-Cys Long was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13571-1gr

 View more versions of this product

Product Code. 15954009

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

SPA-Cys Long Protein
Research Use Only
spa
Other
Untagged
SPA protein was lyophilized with no additives.
1 g
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
AQHDEAQQNA FYQVLNMPNL NADQRNGFIQ SLKDDPSQSA NVLGEAQKLN DSQAPKADAQ QNNFNKDQQS AFYEILNMPN LNEAQRNGFI QSLKDDPSQS TNVLGEAKKL NESQAPKADN NFNKEQQNAF YEILNMPNLN EEQRNGFIQS LKDDPSQSAN LLSEAKKLNE SQAPKADNKF NKEQQNAFYE ILHLPNLNEE QRNGFIQSLK DDPSQSANLL AEAKKLNDAQ APKADNKFNK EQQNAFYEIL HLPNLTEEQR NGFIQSLKDD PSVSKEILAE AKKLNDAQAP KEEDNKKPGK EDGNKPGKED GNKPGKEDNK KPGKEDGNKP GKEDNNKPGK EDGNKPGKED NNKPGKEDGN KPGKEDGNKP GKEDGNGVHV VKPGDTVNDI AKANGTTADK IAADNKLADK NMIKPGQELV VDC
Greater than 97.0% as determined by analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers