missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other SIV p55 Protein

A cDNA sequence encoding the SIV p55 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13512-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953809

  • £1985.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SIV p55 Protein
Research Use Only
Recombinant Protein
Baculovirus
SIV p55 Protein solution containing 10mM Tris, pH 8.0, 140mM NaCl and 400mM L-Arginine.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers