missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other S. Pyogenes arcA Protein
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
Specifications
Specifications
| Name | S. Pyogenes arcA Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | His |
| Buffer | S. Pyogenes arcA is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |
| Purity or Quality Grade | Greater than 80.0% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction