missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Protein-A/G Cys Protein

A cDNA sequence encoding the Protein-A/G Cys was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13164-10mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952699

  • £155.00 / 10mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Protein-A/G Cys Protein
Research Use Only
Recombinant Protein
E. coli
NAAQHDEAQQ NAFYQVLNMP NLNADQRNGF IQSLKDDPSQ SANVLGEAQK LNDSQAPKAD AQQNNFNKDQ QSAFYEILNM PNLNEAQRNG FIQSLKDDPS QSTNVLGEAK KLNESQAPKA DNNFNKEQQN AFYEILNMPN LNEEQRNGFI QSLKDDPSQS ANLLSEAKKL NESQAPKADN KFNKEQQNAF YEILHLPNLN EEQRNGFIQS LKDDPSQSAN LLAEAKKLND AQAPKADNKF NKEQQNAFYE ILHLPNLTEE QRNGFIQSLK DDPSVSKEIL AEAKKLNDAQ APKEEDSLEG SGSGTYKLIL NGKTLKGETT TEAVDAATAE KVFKQYANDN GVDGEWTYDD ATKTFTVTEK PEVIDASELT PAVTTYKLVI NGKTLKGETT TKAVDAETAE KAFKQYANDN GVDGVWTYDD ATKTFTVTEC
Greater than 96.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE.
10 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Protein-A/G was lyophilized without any additives.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers