missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other OmpA S. Enteritidis Protein
A cDNA sequence encoding the OmpA S. Enteritidis was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | OmpA S. Enteritidis Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15921979
|
enQuireBio™
QP12927-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15941979
|
enQuireBio™
QP12927-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931979
|
enQuireBio™
QP12927-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
OmpA S. Enteritidis Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PBS. |