missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other NAF Protein
Recombinant Protein
£283.00 - £835.00
Specifications
Name | NAF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971529
|
enQuireBio™
QP12784-8MG |
8 mg |
£283.00
8mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951529
|
enQuireBio™
QP12784-16MG |
16 mg |
£505.00
16mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961529
|
enQuireBio™
QP12784-32MG |
32 mg |
£835.00
32mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NAF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with no additives. |