missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other MoeX Protein

A cDNA sequence encoding the MoeX was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12716-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951319

  • £660.00 / 500µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MoeX Protein
Research Use Only
Recombinant Protein
E. coli
PBS.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers