missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Malaria Pf. MSP1 Protein
A cDNA sequence encoding the Malaria Pf. MSP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12631-500ug
Additional Details : Weight : 0.01000kg
Specifications
Malaria Pf. MSP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing Phosphate buffered saline and 0.05% NaN3. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |