missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lysostaphin Protein
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Name | Lysostaphin Protein |
| Quantity | 10 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | lss |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MKKTKNNYYT RPLAIGLSTF ALASIVYGGI QNETHASEKS NMDVSKKVAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKAPV ENTAEVETSK APVENTAEVE TSKAPVENTA EVETSKAPVE NTAEVETSKA PVENTAEVET SKAPVENTAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKALV QNRTALRAAT HEHSAQWLNN YKKGYGYGPY PLGINGGMHY GVDFFMNIGT PVKAISSGKI VEAGWSNYGG GNQIGLIEND GVHRQWYMHL SKYNVKVGDY VKAGQIIGWS GSTGYSTAPH LHFQRMVNSF SNSTAQDPMP FLKSAGYGKA GGTVTPTPNT GWKTNKYGTL YKSESASFTP NTDIITRTTG PFRSMPQSGV LKAGQTIHYD EVMKQDGHVW VGYTGNSGQR IYLPVRTWNK STNTLGVLWG TIK |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction