missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Lassa GP1 Protein
A cDNA sequence encoding the Lassa GP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12539-1mg
Additional Details : Weight : 0.01000kg
Specifications
Lassa GP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lassa GP1 protein solution contains Phosphate buffered saline and 100mM arginine and 3M Urea. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Lassa GP1 is >90% pure as determined by 10% PAGE (coomassie staining). |