missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other JEV Protein

A cDNA sequence encoding the JEV was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12471-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15950529

  • £660.00 / 500µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

JEV Protein
Research Use Only
Recombinant Protein
E. coli
20mM Tris-MES pH 6.5, 8M urea, 200mM NaCl and 0.05% Tween-20.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Encephalitis protein is >90% pure as determined by SDS-PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers