missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HPSE Active Protein
Recombinant Protein
£99.15 - £535.00
Specifications
Name | HPSE Active Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The specific activity of Heparanase Active Enzyme in-house standard is about 0.7 Units (1 unit = 1 umole of reducing ends of heparan sulfate substrate formed per minute per mg Heparanase Active Enzyme at 37C). The enzymatic activity of each Heparanase Active Enzyme batch is comparable to the standard as determined by activity assay in which immobilized heparan, released due to heparanase activity, is quantified colorimetrically. Recommended reaction buffer: 20 mM Citrate Phosphate buffer, pH 5.4; 50mM NaCl; 1mM CaCl2. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954588
|
enQuireBio™
QP10668-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964588
|
enQuireBio™
QP10668-4UG |
4 μg |
£164.00
4µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944588
|
enQuireBio™
QP10668-16UG |
16 μg |
£535.00
16µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HPSE Active Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Untagged | |
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The specific activity of Heparanase Active Enzyme in-house standard is about 0.7 Units (1 unit = 1 umole of reducing ends of heparan sulfate substrate formed per minute per mg Heparanase Active Enzyme at 37C). The enzymatic activity of each Heparanase Active Enzyme batch is comparable to the standard as determined by activity assay in which immobilized heparan, released due to heparanase activity, is quantified colorimetrically. Recommended reaction buffer: 20 mM Citrate Phosphate buffer, pH 5.4; 50mM NaCl; 1mM CaCl2. | |
Other | |
rate Phosphate buffer, pH 5.4; 50mM NaCl; 1mM CaCl2. |