missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Hirudin Protein

Product Code. 15984578
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15984578

Brand: enQuireBio™ QP1066610ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Hirudin was constructed and used to recombinantly synthesize the protein.

Specifications

Name Hirudin Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The specific activity was found to be >14,000ATU/mg.
Product Type Recombinant Protein
Cross Reactivity Other
Species P. pastoris
Protein Tag Untagged
Buffer Each mg of protein was lyophilized from a sterile solution containing 20mM PBS pH 7 and 2% mannitol.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.