missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other HEV Mosaic Protein

A cDNA sequence encoding the HEV Mosaic was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12221-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15969738

  • £1100.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HEV Mosaic Protein
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl, 1mM EDTA, 0.5M urea and 50% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
HEV Mosaic protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers