missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HCV NS5A Protein
A cDNA sequence encoding the HCV NS5A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12199-2ug
Additional Details : Weight : 0.01000kg
Specifications
HCV NS5A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSLKATCTAN HDSPDAELIE ANLLWRQEMG GNITRVESEN KVVILDSFDP LVAEEDEREV SVPAEILRKS RRFARALPVW ARPDYNPPLV ETWKKPDY | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The HCV NS5A protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 1mM DTT. |