missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other HCV NS5A Protein

A cDNA sequence encoding the HCV NS5A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12199-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979668

  • £155.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HCV NS5A Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSLKATCTAN HDSPDAELIE ANLLWRQEMG GNITRVESEN KVVILDSFDP LVAEEDEREV SVPAEILRKS RRFARALPVW ARPDYNPPLV ETWKKPDY
Greater than 90% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
The HCV NS5A protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers