missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other hCG Protein
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
Specifications
Specifications
| Name | hCG Protein |
| Quantity | 5 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | The activity was found to be 5625IU/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | Non-species Specific |
| Protein Tag | Untagged |
| Buffer | The hCG was lyophilized with no additives. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction