missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GroEL Protein
A cDNA sequence encoding the GroEL was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12051-1mg
Additional Details : Weight : 0.01000kg
Specifications
GroEL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAAKDVKFGN DARVKMLRGV NVLADAVKVT LGPKGRNVVL DKSFGAPTIT KDGVSVAREIELEDKFENMG AQMVKEVASK ANDAAGDGTT TATVLAQAII TEGLKAVAAG MNPMDLKRGIDKAVTAAVEE LKALSVPCSD SKAIAQVGTI SANSDETVGK LIAEAMDKVG KEGVITVEDGTGLQDELDVV EGMQFDRGYL SPYFINKPET GAVELESPFI LLADKKISNI REMLPVLEAVAKAGKPLLII AEDVEGEALA TAVVNTIRGI VKVAAVKAPG FGDRRKAMLQ DIATLTGGTVISEEIGMELE KATLEDLGQA KRVVINKDTT TIIDGVGEEA AIQGRVAQIR QQIEEATSDYDREKLQERVA KLAGGVAVIK VGAATEVEMK EKKARVEDAL HATRAAVEEG VVAGGGVALIRVASKLADLR GQNEDQNVGI KVALRAMEAP LRQIVLNCGE EPSVVANTVK GGDGNYGYNAATEEYGNMID MGILDPTKVT RSALQYAASV AGLMITTECM VTDLPKNDAA DLGAAGGMGG MGGMGGMM | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The groEL protein contains 25mM Tris buffer (pH 7.5), 100mM NaCl, 5mM DTT and 10% Glycerol. |