missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other GAGA-POZ Protein

A cDNA sequence encoding the GAGA-POZ was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11922-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928808

  • £99.15 / 10µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GAGA-POZ Protein
Research Use Only
Recombinant Protein
E. coli
MSLPMNSLYS LTWGDYGTSL VSAIQLLRCH GDLVDCTLAA GGRSFPAHKI VLCAASPFLLDLLKNTPCKH PVVMLAGVNA NDLEALLEFV YRGEVSVDHA QLPSLLQAAQ CLNIQGLAPQTVTKDDYTTH
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The protein containing 10mM HEPES (pH 7.4) and 25mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers