missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DsbG Protein
A cDNA sequence encoding the DsbG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11706-5ug
Additional Details : Weight : 0.01000kg
Specifications
DsbG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEELPAPVKA IEKQGITIIK TFDAPGGMKG YLGKYQDMGV TIYLTPDGKH AISGYMYNEK GENLSNTLIE KEIYAPAGRE MWQRMEQSHW LLDGKKDAPV IVYVFADPFC PYCKQFWQQA RPWVDSGKVQ LRTLLVGVIK PESPATAAAI LASKDPAKTW QQYEASGGKL KLNVPANVST EQMKVLSDNE KLMDDLGANV TPAIYYMSKE NTLQQAVGLP DQKTLNIIMG NK | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The DsbG protein solution contains 20mM Tris-HCl, pH 8.0, 2mM EDTA and 10% Glycerol. |