missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other DMGO Protein
A cDNA sequence encoding the DMGO was constructed and used to recombinantly synthesize the protein.
£99.15 - £1985.00
Specifications
Name | DMGO Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927998
|
enQuireBio™
QP11673-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947998
|
enQuireBio™
QP11673-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937998
|
enQuireBio™
QP11673-1MG |
1 mg |
£1985.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DMGO Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The Recombinant Dimethylglycine Oxidase solution contains 20mM Tris-HCl pH 7.5 and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MASTPRIVII GAGIVGTNLA DELVTRGWNN ITVLDQGPLN MPGGSTSHAP GLVFQTNPSK TMASFAKYTVEKLLSLTEDG VSCFNQVGGL EVATTETRLA DLKRKLGYAA AWGIEGRLLS PAECQELYPL LDGENILGGL HVPSDGLASA ARAVQLLIKRTESAGVTYRG STTVTGIEQS GGRVTGVQTA DGVIPADIVV SCAGFWGAKI GAMIGMAVPL LPLAHQYVKT TPVPAQQGRN DQPNGARLPILRHQDQDLYY REHGDRYGIG SYAHRPMPVD VDTLGAYAPE TVSEHHMPSR LDFTLEDFLP AWEATKQLLP ALADSEIEDG FNGIFSFTPDGGPLLGESKE LDGFYVAEAV WVTHSAGVAK AMAELLTTGR SETDLGECDI TRFEDVQLTP EYVSETSQQN FVEIYDVLHP LQPRLSPRNLRVSPFHARHK ELGAFFLEAG GWERPYWFEA NAALLKEMPA EWLPPARDAW SGMFSSPIAA AEAWKTRTAV AMYDMTPLKR LEVSGPGALKLLQELTTADL AKKPGAVTYT LLLDHAGGVR SDITVARLSE DTFQLGANGN IDTAYFERAA RHQTQSGSAT DWVQVRDTTG GTCCIGLWGPLARDLVSKVS DDDFTNDGLK YFRAKNVVIG GIPVTAMRLS YVGELGWELY TSADNGQRLW DALWQAGQPF GVIAAGRAAF SSLRLEKGYRSWGTDMTTEH DPFEAGLGFA VKMAKESFIG KGALEGRTEE ASARRLRCLT IDDGRSIVLG KEPVFYKEQA VGYVTSAAYG YTVAKPIAYSYLPGTVSVGD SVDIEYFGRR ITATVTEDPL YDPKMTRLRG | |
Greater than 95.0% as determined by SDS-PAGE. |