missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Der F1 Protein

A cDNA sequence encoding the Der F1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11656-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937938

  • £660.00 / 500µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Der F1 Protein
Research Use Only
DERF1
Other
His
60mM NaCl, 50mM Tris-HCl pH 8.0 and 1.2M Urea.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MKFVLAIASL LVLSTVYARP ASIKTFEEFK KAFNKNYATV EEEEVARKNF LESLKYVEAN KGAINHLSDL SLDEFKNRYL MSAEAFEQLK TQFDLNAETS ACRINSVNVP SELDLRSLRT VTPIRMQGGC GSCWAFSGVA ATESAYLAYR NTSLDLSEQE LVDCASQHGC HGDTIPRGIE YIQQNGVVEE RSYPYVAREQ RCRRPNSQHY GISNYCQIYP PDVKQIREAL TQTHTAIAVI IGIKDLRAFQ HYDGRTIIQH DNGYQPNYHA VNIVGYGSTQ GDDYWIVRNS WDTTWGDSGY GYFQAGNNLM MIEQYPYVVI M
Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers