missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Carbonic Anhydrase II Protein

A cDNA sequence encoding the Carbonic Anhydrase II was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11275-1mg

 View more versions of this product

Product Code. 15956688

  • £3085.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

944832
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MKDIDTLISN NALWSKMLVE EDPGFFEKLAQAQKPRFLWI GCSDSRVPAE RLTGLEPGEL FVHRNVANLV IHTDLNCLSV VQYAVDVLEV EHIIICGHYG CGGVQAAVEN PELGLINNWL HIRDIWFKH SSLLGEMPQE RRLDTLCELN VMEQVYNLGH STIMQSAWKR GQKVTIHGWA YGIHDGLLRD LDVTATNRET LEQRYRHGIS NLKLKHANHK
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Carbonic Anhydrase II Protein
Research Use Only
can
Other
His
The Carbonic Anhydrase 2 enzyme is supplied in 20mM Tris pH 8 and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers