missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia p100 Protein

A cDNA sequence encoding the Borrelia p100 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11191-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956428

  • £155.00 / 20µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Borrelia p100 Protein
Research Use Only
Recombinant Protein
Baculovirus
Borrelia p100 (0.77 mg/1ml) is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers