missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Borrelia p100 Protein
A cDNA sequence encoding the Borrelia p100 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11191-1mg
Additional Details : Weight : 0.01000kg
Specifications
Borrelia p100 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
Borrelia p100 (0.77 mg/1ml) is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol. |