missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia OspC Protein

A cDNA sequence encoding the Borrelia OspC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11190-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936428

  • £99.15 / 5µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Borrelia OspC Protein
Research Use Only
Recombinant Protein
E. coli
Borrelia OspC (1.74 mg/1ml) is supplied in 16mM HEPES buffer pH 7.0, 300mM NaCl and 20% glycerol.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers