missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia Miyamotoi GlpQ Protein

A cDNA sequence encoding the Borrelia Miyamotoi GlpQ was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11187-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15926418

  • £3965.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Borrelia Miyamotoi GlpQ Protein
Research Use Only
Recombinant Protein
E. coli
Borrelia Miyamotoi GlpQ is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers