missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia Garinii OspC Protein

A cDNA sequence encoding the Borrelia Garinii OspC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11184-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956408

  • £99.15 / 5µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Borrelia Garinii OspC Protein
Research Use Only
Recombinant Protein
E. coli
Borrelia Garinii OspC is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers