missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Borrelia BmpA Protein

A cDNA sequence encoding the Borrelia BmpA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11181-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946398

  • £3965.00 / 1mg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Borrelia BmpA Protein
Research Use Only
Recombinant Protein
E. coli
Borrelia BmpA (1.3 mg/1ml) is supplied in 20mM HEPES buffer pH 8.0, 6M Urea and 200mM NaCl.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Greater than 80.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers