missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other B. Microti p32 Protein
A cDNA sequence encoding the B. Microti p32 was constructed and used to recombinantly synthesize the protein.
£770.00 - £4955.00
Specifications
Name | B. Microti p32 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946198
|
enQuireBio™
QP11116-50UG |
50 μg |
£770.00
50µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926198
|
enQuireBio™
QP11116-100UG |
100 μg |
£1100.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936198
|
enQuireBio™
QP11116-1MG |
1 mg |
£4955.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
B. Microti p32 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Greater than 80.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
B. Microti p32 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |