missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Angiostatin K1-4 Protein

Product Code. 15934128
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15934128

Brand: enQuireBio™ QP105212ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Angiostatin K1-4 was constructed and used to recombinantly synthesize the protein.

Specifications

Name Angiostatin K1-4 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Human Angiostatin Kringles 1-4 significantly inhibits basic-FGF induced endothelial cell proliferation and migration at concentration ranging from 300nM-1.0 uM.
Product Type Recombinant Protein
Cross Reactivity Other
Species Non-species Specific
Protein Tag Untagged
Buffer Lyophilized from a (1 mg/ml) solution in containing 20mM Hepes buffer pH 8.2 and 20mM NaCl.
Purity or Quality Grade Greater than 98.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.