missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Aln G 4.0101 Protein
A cDNA sequence encoding the Aln G 4.0101 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11008-100ug
Additional Details : Weight : 0.01000kg
Specifications
Aln G 4.0101 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MADDHPQDQA EHERIFKCFD ANGDGKISAS ELGDALKTLG SVTPDEVKHM MAEIDTDGDG FISFQEFTNF ARANRGLVKD VAKIF | |
Greater than 80.0% as determined by SDS-PAGE. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Aln G 4.0101 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |