missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Oropouche virus nucleoProtein
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
Specifications
Specifications
| Name | Oropouche virus nucleoProtein |
| Quantity | 500 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Untested |
| Product Type | Recombinant Protein |
| Cross Reactivity | Oropouche virus, strain TRVL9760 |
| Species | E. coli |
| Protein Tag | Native |
| Buffer | 15mM Tris-Hcl, 225mM NaCl, 20% glycerol, pH7.5. |
| Purity or Quality Grade | >90% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction