Learn More
enQuireBio™ Recombinant Multi Species ACTH Protein
Recombinant Protein
Brand: enQuireBio™ QP10950-2mg
Additional Details : Weight : 0.01000kg
Specifications
Multi Species ACTH Protein | |
Research Use Only | |
Recombinant Protein | |
Untagged | |
The ACTH hormone was lyophilized with no additives. |
2 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Multi Species | |
Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |