missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse YM1/Chitinase 3-like 3 Protein

Product Code. 18318917
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18318917 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18318917 Supplier R&D Systems™ Supplier No. 2446CH050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of FaDu human squamous cell carcinoma cells. When 5 x 104 cells/well are added to Recombinant Mouse YM1/Chitinase 3-like 3 coated plates, cell adhesion is enhanced in a dose dependent manner after 1 hour incubation at 37 °C. The ED50 for this effect is 0.3-1.2 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_034022
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1117
Molecular Weight (g/mol) M.W. (Observed): 43 kDa, reducing conditions; M.W. (theoretical): 43 kDa
Name Chitinase 3-like 3/ECF-L
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse YM1/Chitinase 3-like 3 protein Met1-Tyr398, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Chil3
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein YM1/Chitinase 3-like 3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.