missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Wnt-10b Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£406.04
Specifications
Gene ID (Entrez) | 22410 |
---|---|
Molecular Weight (g/mol) | 40.1 kDa |
Name | Wnt-10b |
Quantity | 10μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-10b protein Met1-Lys389 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18166160
|
R&D Systems™
2110-WN-010 |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
£406.04
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18136970
|
R&D Systems™
2110-WN-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. |
£406.04
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse Wnt-10b Protein is derived from CHO. The Recombinant Mouse Wnt-10b Protein has been validated for the following applications: Bioactivity.Specifications
22410 | |
Wnt-10b | |
Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-10b protein Met1-Lys389 | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Unconjugated | |
85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
40.1 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, Wnt10b, WNT12, WNT-12 | |
Recombinant |