missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse VEGFR1/Flt-1 Fc Chimera Protein
Description
The Recombinant Mouse VEGFR1/Flt-1 Fc Chimera Protein is derived from NS0. The Recombinant Mouse VEGFR1/Flt-1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS. |
| Gene ID (Entrez) | 14254 |
| Molecular Weight (g/mol) | 110 kDa (monomer) |
| Name | VEGF R1/Flt-1 Fc Chimera |
| Quantity | 100μg |
| Source | Mouse myeloma cell line,NS0-derived mouse VEGFR1/Flt-1 protein, (N-terminus) Mouse VEGFR1 (Ser27-Glu759) Accession &Num; P35969 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?