missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse VEGF120 Protein

Product Code. p-200083622 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
This item is not returnable. View return policy

Product Code. 18671541

Brand: Novus Biologicals™ NBP276334500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. The ED50 for this effect is 1-4 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) MolecularWeight-theroretical: 20.7 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of VEGF120 as determined by LAL method.
Gene Symbol VEGFA
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Inhibitor Properties As a result of glycosylation, it migrates to at least two bands with molecular weights ranging from 20.7 kDa in SDS-PAGE under reducing conditions.
Purity or Quality Grade >95%, by SDS-PAGE
Protein VEGF120
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.