missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed)

Product Code. 18176600
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18176600 Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA with BSA as a carrier protein. 10µg
18359227 Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18176600 Supplier R&D Systems™ Supplier No. 462TEC010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed) is derived from E. coli. The Recombinant Mouse TRANCE/RANK L/TNFSF11 (E. coli-expressed) has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 21943
Molecular Weight (g/mol) 19 kDa
Name TRANCE/TNFSF11/RANK L
Quantity 10 μg
Source E. coli-derived mouse TRANCE/TNFSF11/RANK L protein Lys158-Asp316,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.