missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TRAIL/TNFSF10 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 with Trehalose and with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 .
Molecular Weight (g/mol):
20 kDa (monomer)
20 kda
Unit Size:
10µg
Description
The Recombinant Mouse TRAIL/TNFSF10 Protein is derived from E. coli. The Recombinant Mouse TRAIL/TNFSF10 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 with Trehalose and with BSA as a carrier protein. |
| Gene ID (Entrez) | 22035 |
| Molecular Weight (g/mol) | 20 kda |
| Name | TRAIL/TNFSF10 |
| Quantity | 10 μg |
| Source | E. coli-derived mouse TRAIL/TNFSF10 protein Pro118-Asn291 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.01 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction