missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TRAIL Protein

A cDNA sequence encoding the TRAIL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10892-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945488

  • £1495.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

22035
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Fully biologically active when compared to standard. The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2,000,000 IU/mg in the presence of actinomycin D.
Mouse
Untagged
The protein was lyophilized containing PBS, pH 7.4, and 3mM DTT.
TRAIL Protein
Research Use Only
Tnfsf10
Recombinant Protein
E. coli
MPRGGRPQKV AAHITGITRR SNSALIPISK DGKTLGQKIE SWESSRKGHS FLNHVLFRNG ELVIEQEGLY YIYSQTYFRF QEAEDASKMV SKDKVRTKQL VQYIYKYTSY PDPIVLMKSA RNSCWSRDAE YGLYSIYQGG LFELKKNDRI FVSVTNEHLM DLDQEASFFG AFLIN
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers