missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse TNF RI/TNFRSF1A Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
TNF Receptor I Protein is fully biologically active when compared to standard. The ED50 as determined by its ability to inhibit the TNF-alpha mediated cytotoxicity in the L-929 cells is less than 1 μg/mL, corresponding to a specific activity of >1000 IU/mg in the presence of 0.1 ng/mL of rMuTNF-alpha.
Specifications
Specifications
| Accession Number | P25118, P25118, P25118 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 7132 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 21.1 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of TNF RI/TNFRSF1A as determined by LAL method. |
| Gene Alias | CD120a, CD120a antigen, FPF, p55, p55-R, p60, sTNF RI, sTNF RI/TNFRSF1A, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor |
| Gene Symbol | TNFRSF1A |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction