missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TLR12 Fc Chimera Protein
Click to view available options
:
25ug; Carrier Free
Description
The Recombinant Mouse TLR12 Fc Chimera Protein is derived from CHO. The Recombinant Mouse TLR12 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 384059 |
| Molecular Weight (g/mol) | 102 kDa (monomer) |
| Name | TLR12 Fc Chimera |
| Quantity | 25μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse TLR12 protein, (N-terminus) Mouse TLR12 (Thr22-Lys709) Accession &Num; NP_991392 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction