missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TIM-1/KIM-1/HAVCR Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
Unit Size:
50µg
Description
The Recombinant Mouse TIM-1/KIM-1/HAVCR Protein is derived from NS0. The Recombinant Mouse TIM-1/KIM-1/HAVCR Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. |
| Gene ID (Entrez) | 171283 |
| Molecular Weight (g/mol) | 21.8 kDa |
| Name | TIM-1/KIM-1/HAVCR |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse TIM-1/KIM-1/HAVCR protein Tyr22-Thr212,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | CD365, HAVcr-1, HAVCR1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction