missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Thrombospondin-1 Protein

Product Code. 18157320
Click to view available options
:
50ug; Carrier Free
This item is not returnable. View return policy

Product Code. 18157320

Brand: R&D Systems™ 7859TH050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of SVEC4-10 mouse vascular endothelial cells. The ED50 for this effect is 0.1-0.5 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_035710
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 7057
Molecular Weight (g/mol) M.W. (Observed): 130-155 kDa, reducing conditions; M.W. (theoretical): 129 kDa
Name Thrombospondin-1
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse Thrombospondin-1 protein Mouse Thrombospondin-1 (Asn19-Ser1171) (N-terminus) Accession # NP_035710 GGGSGGGSGGGS 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Thbs1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Thrombospondin-1
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis