missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thrombopoietin / THPO Protein
Recombinant Protein
£2755.00 - £4515.00
Specifications
Accession Number | NP_001166976.1 |
---|---|
Gene ID (Entrez) | 21832 |
Name | Thrombopoietin / THPO Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956129
|
enQuireBio™
QP5439-500UG |
500 μg |
£2755.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946129
|
enQuireBio™
QP5439-1MG |
1 mg |
£4515.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001166976.1 | |
Thrombopoietin / THPO Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
MO7e cell proliferation significantly induced by less than 5 ng/ml (activity of at least 2.0 x 10^5 units/mg). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
21832 | |
Research Use Only | |
THPO | |
Recombinant Protein | |
E. coli | |
MSPVAPACDP RLLNKLLRDS HLLHSRLSQC PDVDPLSIPV LLPAVDFSLG EWKTQTEQSK AQDILGAVSL LLEGVMAARG QLEPSCLSSL LGQLSGQVRL LLGALQGLLG TQLPLQGRTT AHKDPNALFL SLQQLLRGKV RFLLLVEGPT LCVRRTLPTT AVPSSTSQLL TLNKF | |
Greater than 95% as determined by SDS-PAGE. |