missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Thioredoxin domain-containing protein 12 Protein

A DNA sequence encoding the Mus musculus (Mouse) Thioredoxin domain-containing protein 12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8212-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083301

  • £560.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_079610.1
Thioredoxin domain-containing protein 12 Protein
Research Use Only
TXNDC12
Recombinant Protein
E. coli
RTGLGKGFGD HIHWRTLEDG KKEAAASGLP LMVIIHKSWC GACKALKPKF AESTEISELS HNFVMVNLED EEEPRDEDFS PDGGYIPRIL FLDPSGKVRP EIINESGNPS YKYFYVSAEQ VVQGMKEAQE RLTGDAFREK HFQDEL
Greater than 90% as determined by SDS-PAGE.
66073
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers