missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse Testican 3/SPOCK3 His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 22-436 of Mouse Testican 3/SPOCK3 The Recombinant Mouse Testican 3/SPOCK3 Protein is derived from Baculovirus. The Recombinant Mouse Testican 3/SPOCK3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 0.25mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | In PBS (pH 7.4) containing 10% glycerol |
| Gene ID (Entrez) | 50859 |
| Molecular Weight (g/mol) | TMW: 47.9kDa |
| Quantity | 20 μg |
| Source | Baculovirus |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −70°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | CWCV, and Kazal-like domains proteoglycan 3, HSAJ1454, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3, TES-3, testican 3, TICN3 |
| Common Name | Testican 3/SPOCK3 |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction