missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TEK Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | TEK Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Species | Baculovirus |
| Protein Tag | His |
| Sequence | AMDLILINSL PLVSDAETSL TCIASGWHPH EPITIGRDFE ALMNQHQDPL EVTQDVTREW AKKVVWKREK ASKINGAYFC EGRVRGQAIR IRTMKMRQQA SFLPATLTMT VDRGDNVNIS FKKVLIKEED AVIYKNGSFI HSVPRHEVPD ILEVHLPHAQ PQDAGVYSAR YIGGNLFTSA FTRLIVRRCE AQKWGPDCSR PCTTCKNNGV CHEDTGECIC PPGFMGRTCE KACEPHTFGR TCKERCSGPE GCKSYVFCLP DPYGCSCATG WRGLQCNEAC PSGYYGPDCK LRCHCTNEEI CDRFQGCLCS QGWQGLQCEK EGRPRMTPQI EDLPDHIEVN SGKFNPICKA SGWPLPTSEE MTLVKPDGTV LQPNDFNYTD RFSVAIFTVN RVLPPDSGVW VCSVNTVAGM VEKPFNISVK VLPEPLHAPN VIDTGHNFAI INISSEPYFG DGPIKSKKLF YKPVNQAWKY IEVTNEIFTL NYLEPRTDYE LCVQLARPGE GGEGHPGPVR RFTTASIGLP PPRGLSLLPK SQTALNLTWQ PIFTNSEDEF YVEVERRSLQ TTSDQQNIKV PGNLTSVLLS NLVPREQYTV RARVNTKAQG EWSEELRAWT LSDILPPQPE NIKISNITDS TAMVSWTIVD GYSISSIIIR YKVQGKNEDQ HIDVKIKNAT VTQYQLKGLE PETTYHVDIF AENNIGSSNP AFSHELRTLP HSPASADLGG GKMLHHHHHH |
| Buffer | TEK protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction