missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse ST6GALNAC2 Protein
Description
Measured by its ability to transfer Neu5Ac from CMP-Neu5Ac to fetuin of fetal calf serum. The specific activity is >700 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P70277 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 10610 |
| Molecular Weight (g/mol) | M.W. (Observed): 40-50 kDa, reducing conditions; M.W. (theoretical): 40 kDa |
| Name | ST6 Sialyltransferase 2/ST6GALNAC2 |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse ST6 Sialyltransferase 2/ST6GALNAC2 protein Ser28-Arg373, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?